Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ecscr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772057.1 | XP_008770279.1 | 4 - 5 | 356 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |