Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ecscr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772057.1 | XP_008770279.1 | 6 - 7 | 566 | 65 | |
XM_008772058.1 | XP_008770280.1 | 5 - 6 | 521 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |