Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dyrk1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012791.2 | NP_036923.1 | 9 - 10 | 1373 | 100 | |
XM_006248031.2 | XP_006248093.1 | 9 - 10 | 4318 | 100 | ||
XM_008768562.1 | XP_008766784.1 | 8 - 9 | 3581 | 100 | ||
XM_008768563.1 | XP_008766785.1 | 10 - 11 | 4454 | 100 | ||
XM_008768564.1 | XP_008766786.1 | 9 - 10 | 1420 | 100 | ||
XM_008768565.1 | XP_008766787.1 | 9 - 10 | 1495 | 100 | ||
GenBank mRNA | X79769.1 | - | 9 - 10 | 1375 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Rattus norvegicus Probe spans exons |