Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017177.1 | NP_058873.1 | 5 - 6 | 954 | 92 | |
XM_006242181.2 | XP_006242243.2 | 5 - 6 | 1620 | 92 | ||
XM_008765677.1 | XP_008763899.1 | 5 - 6 | 1617 | 92 | ||
GenBank mRNA | AB006607.1 | - | 5 - 6 | 954 | 92 | |
BC060515.1 | - | 5 - 6 | 930 | 5598184 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |