Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabl2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013221.1 | NP_001013239.1 | 4 - 5 | 551 | 57 | |
XM_006242225.2 | XP_006242287.1 | 4 - 5 | 632 | 57 | ||
XM_006242226.2 | XP_006242288.1 | 4 - 5 | 633 | 57 | ||
XM_006242227.1 | XP_006242289.1 | 4 - 5 | 353 | 57 | ||
XM_006242228.2 | XP_006242290.1 | 4 - 5 | 588 | 57 | ||
GenBank mRNA | BC083624.1 | - | 4 - 5 | 551 | 7134121 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |