Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncaph2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024877.1 | NP_001020048.1 | 16 - 17 | 1433 | 75 | |
XM_006242187.2 | XP_006242249.1 | 15 - 16 | 1395 | 75 | ||
GenBank mRNA | BC097379.1 | - | 16 - 17 | 1433 | 7445599 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |