Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncaph2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024877.1 | NP_001020048.1 | 12 - 13 | 1106 | 59 | |
XM_006242186.2 | XP_006242248.1 | 11 - 12 | 1068 | 59 | ||
XM_006242187.2 | XP_006242249.1 | 11 - 12 | 1068 | 59 | ||
XM_006242188.2 | XP_006242250.1 | 11 - 12 | 1029 | 59 | ||
GenBank mRNA | BC097379.1 | - | 12 - 13 | 1106 | 7445599 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |