Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trabd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106788.1 | NP_001100258.1 | 3 - 4 | 217 | 76 | |
XM_006242182.2 | XP_006242244.1 | 3 - 4 | 779 | 76 | ||
XM_006242183.2 | XP_006242245.1 | 3 - 4 | 632 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |