Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trabd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106788.1 | NP_001100258.1 | 7 - 8 | 778 | 73 | |
XM_006242182.2 | XP_006242244.1 | 7 - 8 | 1340 | 73 | ||
XM_006242183.2 | XP_006242245.1 | 7 - 8 | 1193 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |