Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013200.1 | NP_037332.1 | 17 - 18 | 2182 | 83 | |
XM_006242180.2 | XP_006242242.1 | 17 - 18 | 2638 | 83 | ||
GenBank mRNA | BC085761.1 | - | 17 - 18 | 2194 | 7193798 | 83 |
D43623.1 | - | 16 - 17 | 2169 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |