Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prickle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199396.1 | NP_955428.1 | 3 - 4 | 871 | 96 | |
XM_006242213.2 | XP_006242275.1 | 2 - 3 | 304 | 96 | ||
XM_006242214.2 | XP_006242276.1 | 3 - 4 | 2768 | 96 | ||
XM_006242215.2 | XP_006242277.1 | 3 - 4 | 699 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |