Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gxylt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100887.1 | NP_001094357.1 | 5 - 6 | 895 | 75 | |
XM_006242192.2 | XP_006242254.1 | 4 - 5 | 803 | 75 | ||
GenBank mRNA | AY623036.1 | - | 3 - 4 | 498 | 75 | |
BC167068.1 | - | 4 - 5 | 649 | 8363586 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |