Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053447.1 | NP_445899.1 | 4 - 5 | 564 | 87 | |
GenBank mRNA | BC161939.1 | - | 4 - 5 | 539 | 87 | |
DY318702.1 | - | 4 - 5 | 564 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |