Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106391.2 | NP_001099861.1 | 2 - 3 | 176 | 102 | |
GenBank mRNA | BC158719.1 | - | 2 - 3 | 176 | 9089752 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |