Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tube1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108536.1 | NP_001102006.1 | 3 - 4 | 414 | 114 | |
XM_006256528.2 | XP_006256590.1 | 4 - 5 | 332 | 114 | ||
XM_006256529.1 | XP_006256591.1 | 3 - 4 | 202 | 114 | ||
GenBank mRNA | BC158720.1 | - | 4 - 5 | 316 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |