Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tube1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108536.1 | NP_001102006.1 | 5 - 6 | 654 | 85 | |
XM_006256528.2 | XP_006256590.1 | 6 - 7 | 572 | 85 | ||
XM_006256529.1 | XP_006256591.1 | 5 - 6 | 442 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |