Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tube1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256528.2 | XP_006256590.1 | 1 - 2 | 151 | 68 | |
GenBank mRNA | BC158720.1 | - | 1 - 2 | 135 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |