Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170557.1 | NP_001164028.1 | 5 - 6 | 658 | 150 | |
XM_006240921.2 | XP_006240983.1 | 5 - 6 | 612 | 150 | ||
XM_006240922.2 | XP_006240984.1 | 5 - 6 | 612 | 150 | ||
XM_006240923.2 | XP_006240985.1 | 4 - 5 | 495 | 150 | ||
XM_006240925.2 | XP_006240987.1 | 5 - 6 | 612 | 150 | ||
XM_006240926.2 | XP_006240988.1 | 5 - 6 | 612 | 150 | ||
XM_008765098.1 | XP_008763320.1 | 5 - 6 | 612 | 150 | ||
XM_008765099.1 | XP_008763321.1 | 6 - 7 | 584 | 150 | ||
XM_008765100.1 | XP_008763322.1 | 5 - 6 | 742 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |