Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031649.1 | NP_001026819.1 | 3 - 4 | 411 | 112 | |
XM_006240896.1 | XP_006240958.1 | 4 - 5 | 568 | 112 | ||
XM_006240897.2 | XP_006240959.1 | 3 - 4 | 1037 | 112 | ||
XM_008765091.1 | XP_008763313.1 | 3 - 4 | 1062 | 112 | ||
XM_008765092.1 | XP_008763314.1 | 4 - 5 | 807 | 112 | ||
GenBank mRNA | BC098923.1 | - | 3 - 4 | 411 | 7461534 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |