Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pias4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100757.1 | NP_001094227.1 | 3 - 4 | 575 | 66 | |
GenBank mRNA | BC169045.1 | - | 3 - 4 | 575 | 9100787 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |