Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mum1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108736.2 | NP_001102206.2 | 3 - 4 | 225 | 145 | |
XM_006240973.2 | XP_006241035.1 | 3 - 4 | 258 | 145 | ||
XM_006240974.2 | XP_006241036.1 | 3 - 4 | 291 | 145 | ||
XM_006240975.2 | XP_006241037.1 | 3 - 4 | 1212 | 145 | ||
GenBank mRNA | BC160828.1 | - | 3 - 4 | 225 | 7453186 | 145 |
CV126168.1 | - | 3 - 4 | 251 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |