Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gzmm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057183.1 | NP_476531.1 | 4 - 5 | 657 | 129 | |
GenBank mRNA | BI278865.1 | - | 1 - 2 | 129 | ||
FQ233430.1 | - | 4 - 5 | 684 | 129 | ||
L05175.1 | - | 4 - 5 | 600 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |