Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gna15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053542.1 | NP_445994.1 | 5 - 6 | 895 | 77 | |
XM_006241021.2 | XP_006241083.1 | 5 - 6 | 1267 | 77 | ||
GenBank mRNA | AB015308.1 | - | 5 - 6 | 970 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |