Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uqcr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126097.2 | NP_001119569.1 | 1 - 2 | 99 | 58 | |
NM_001287109.1 | NP_001274038.1 | 1 - 2 | 99 | 58 | ||
XM_006241008.2 | XP_006241070.1 | 1 - 2 | 138 | 58 | ||
GenBank mRNA | BC157815.1 | - | 1 - 2 | 75 | 8366041 | 58 |
CB783645.1 | - | 1 - 2 | 77 | 58 | ||
FM080527.1 | - | 1 - 2 | 101 | 58 | ||
FQ217807.1 | - | 1 - 2 | 109 | 58 | ||
FQ217927.1 | - | 1 - 2 | 70 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |