Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4f17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191986.1 | NP_001178915.1 | 350 | 133 | ||
XM_006241085.2 | XP_006241147.1 | 1502 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus 1700 Inventoried Probe spans exons |