Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RragB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053972.2 | NP_446424.1 | 6 - 7 | 1115 | 128 | |
XM_006256779.2 | XP_006256841.1 | 5 - 6 | 1129 | 128 | ||
XM_008773066.1 | XP_008771288.1 | 5 - 6 | 1270 | 128 | ||
XM_008773067.1 | XP_008771289.1 | 4 - 5 | 1186 | 128 | ||
GenBank mRNA | BC078760.1 | - | 6 - 7 | 653 | 7123902 | 128 |
BC087698.1 | - | 6 - 7 | 1115 | 7314028 | 128 |
Gene Symbol: LOC100909655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003752022.3 | XP_003752070.1 | 1214 | 128 | ||
XM_006256775.2 | XP_006256837.1 | 1129 | 128 | |||
XM_008773107.1 | XP_008771329.1 | 1270 | 128 | |||
XM_008773108.1 | XP_008771330.1 | 1186 | 128 | |||
XM_008773109.1 | XP_008771331.1 | 5 - 6 | 1270 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |