Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pglyrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191961.1 | NP_001178890.1 | 277 | 81 | ||
XM_006232834.2 | XP_006232896.1 | 2 - 3 | 407 | 81 | ||
XM_006232836.2 | XP_006232898.1 | 2 - 3 | 407 | 81 |
Gene Symbol: Pglyrp3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232841.2 | XP_006232903.1 | 2 - 3 | 407 | 81 | |
XM_006232842.2 | XP_006232904.1 | 2 - 3 | 407 | 81 | ||
XM_006232844.2 | XP_006232906.1 | 2 - 3 | 407 | 81 | ||
XM_008761252.1 | XP_008759474.1 | 2 - 3 | 407 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |