Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107454.1 | NP_001100924.1 | 7 - 8 | 768 | 77 | |
XM_008758691.1 | XP_008756913.1 | 3 - 4 | 1466 | 77 | ||
XM_008758692.1 | XP_008756914.1 | 10 - 11 | 2502 | 77 | ||
XM_008758693.1 | XP_008756915.1 | 7 - 8 | 1283 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Inventoried Probe spans exons |