Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Radil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CV116605.1 | - | 3 - 4 | 505 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus Probe spans exons |