Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsph10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013867.1 | NP_001013889.1 | 14 - 15 | 1945 | 91 | |
XM_006248858.2 | XP_006248920.1 | 15 - 16 | 2772 | 91 | ||
XM_006248859.2 | XP_006248921.1 | 15 - 16 | 2640 | 91 | ||
XM_006248860.2 | XP_006248922.1 | 15 - 16 | 2772 | 91 | ||
XM_006248861.2 | XP_006248923.1 | 14 - 15 | 2681 | 91 | ||
GenBank mRNA | BC081935.1 | - | 14 - 15 | 1945 | 7114952 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |