Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191837.2 | NP_001178766.1 | 10 - 11 | 2942 | 125 | |
XM_006229310.2 | XP_006229372.1 | 9 - 10 | 3787 | 125 | ||
XM_006229313.2 | XP_006229375.1 | 8 - 9 | 3667 | 125 | ||
XM_008759399.1 | XP_008757621.1 | 7 - 8 | 4497 | 125 | ||
XM_008759400.1 | XP_008757622.1 | 9 - 10 | 2428 | 125 | ||
XM_008759401.1 | XP_008757623.1 | 9 - 10 | 3731 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |