Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024370.3 | NP_077346.3 | 4 - 5 | 613 | 86 | |
GenBank mRNA | M81142.1 | - | 4 - 5 | 595 | 86 | |
X63324.1 | - | 669 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Rattus norvegicus Probe spans exons |