Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghrhr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012850.1 | NP_036982.1 | 3 - 4 | 292 | 103 | |
XM_008762872.1 | XP_008761094.1 | 3 - 4 | 1655 | 103 | ||
GenBank mRNA | AF122055.1 | - | 3 - 4 | 265 | 103 | |
L01407.1 | - | 3 - 4 | 292 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |