Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf2bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749741.3 | XP_003749789.1 | 12 - 13 | 1670 | 116 | |
XM_006236474.2 | XP_006236536.1 | 12 - 13 | 1544 | 116 | ||
XM_006236475.2 | XP_006236537.1 | 10 - 11 | 1481 | 116 | ||
XM_008762933.1 | XP_008761155.1 | 12 - 13 | 1305 | 116 | ||
GenBank mRNA | AY724479.1 | - | 12 - 13 | 1634 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |