Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf2bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749741.3 | XP_003749789.1 | 9 - 10 | 1357 | 113 | |
XM_006236474.2 | XP_006236536.1 | 9 - 10 | 1231 | 113 | ||
XM_006236475.2 | XP_006236537.1 | 7 - 8 | 1168 | 113 | ||
XM_008762933.1 | XP_008761155.1 | 9 - 10 | 992 | 113 | ||
GenBank mRNA | AY724479.1 | - | 9 - 10 | 1321 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |