Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109022.1 | NP_001102492.1 | 2 - 3 | 418 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons |