Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017221.1 | NP_058917.1 | 1 - 2 | 613 | 143 | |
GenBank mRNA | L27340.1 | - | 1 - 2 | 613 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Rattus norvegicus Probe spans exons |