Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022392.1 | NP_071787.1 | 2 - 3 | 530 | 63 | |
GenBank mRNA | BC078827.1 | - | 2 - 3 | 540 | 7125208 | 63 |
L13619.1 | - | 2 - 3 | 530 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |