Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tceal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009675.1 | NP_001009675.1 | 1 - 2 | 103 | 99 | |
XM_006257294.1 | XP_006257356.1 | 1 - 2 | 168 | 99 | ||
GenBank mRNA | BC087575.1 | - | 1 - 2 | 103 | 7302183 | 99 |
FQ214955.1 | - | 1 - 2 | 36 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons |