Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm9sf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006231413.1 | XP_006231475.1 | 7 - 8 | 1357 | 120 | |
GenBank mRNA | FQ229341.1 | - | 3 - 4 | 381 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |