Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm9sf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006231413.1 | XP_006231475.1 | 12 - 13 | 1941 | 70 | |
GenBank mRNA | FQ209899.1 | - | 9 - 10 | 1056 | 70 | |
FQ229341.1 | - | 8 - 9 | 965 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |