Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfxn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108527.1 | NP_001101997.1 | 4 - 5 | 306 | 110 | |
XM_006231684.2 | XP_006231746.1 | 3 - 4 | 2367 | 110 | ||
XM_006231685.2 | XP_006231747.1 | 3 - 4 | 1203 | 110 | ||
GenBank mRNA | FQ215895.1 | - | 4 - 5 | 295 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |