Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Habp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001505.1 | NP_001001505.1 | 7 - 8 | 758 | 58 | |
XM_006231631.2 | XP_006231693.1 | 7 - 8 | 806 | 58 | ||
XM_006231632.2 | XP_006231694.1 | 6 - 7 | 695 | 58 | ||
XM_006231633.2 | XP_006231695.1 | 6 - 7 | 692 | 58 | ||
GenBank mRNA | AB177406.1 | - | 7 - 8 | 758 | 58 | |
BC129080.1 | - | 6 - 7 | 644 | 8374257 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |