Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Habp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001505.1 | NP_001001505.1 | 8 - 9 | 854 | 98 | |
XM_006231631.2 | XP_006231693.1 | 8 - 9 | 902 | 98 | ||
XM_006231632.2 | XP_006231694.1 | 7 - 8 | 791 | 98 | ||
XM_006231633.2 | XP_006231695.1 | 7 - 8 | 788 | 98 | ||
GenBank mRNA | AB177406.1 | - | 8 - 9 | 854 | 98 | |
BC129080.1 | - | 7 - 8 | 740 | 8374257 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |