Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdlim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017365.2 | NP_059061.1 | 2 - 3 | 314 | 78 | |
GenBank mRNA | BC072465.1 | - | 2 - 3 | 305 | 7100446 | 78 |
U23769.1 | - | 2 - 3 | 314 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |