Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106364.1 | NP_001099834.1 | 7 - 8 | 817 | 99 | |
XM_006231470.2 | XP_006231532.1 | 7 - 8 | 989 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |