Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aida | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127600.1 | NP_001121072.1 | 3 - 4 | 388 | 62 | |
GenBank mRNA | BC161899.1 | - | 3 - 4 | 388 | 8367117 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |