Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011907.1 | NP_001011907.1 | 5 - 6 | 715 | 113 | |
XM_003751277.2 | XP_003751325.1 | 5 - 6 | 728 | 113 | ||
GenBank mRNA | BC082067.1 | - | 5 - 6 | 715 | 7132777 | 113 |
FQ215922.1 | - | 5 - 6 | 719 | 113 |
Gene Symbol: LOC103690046 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769779.1 | XP_008768001.1 | 2 - 3 | 346 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |