Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomm40l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083338.1 | NP_001076807.1 | 4 - 5 | 382 | 89 | |
XM_006250247.2 | XP_006250309.1 | 5 - 6 | 861 | 89 | ||
GenBank mRNA | AB274732.1 | - | 4 - 5 | 382 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |