Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casq1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159594.1 | NP_001153066.1 | 1 - 2 | 446 | 88 | |
GenBank mRNA | CA334109.1 | - | 1 - 2 | 446 | 88 | |
FQ224875.1 | - | 2 - 3 | 273 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |